missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(2-Phenoxyethyl)thio]acetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Linear Formula | C6H5O(CH2)2SCH2CO2H |
Quantity | 25 g |
Formula Weight | 212.27 |
Percent Purity | 97% |
Chemical Name or Material | 2-[(2-Phenoxyethyl)thio]acetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'