missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96 Well 2.2mL Polypropylene Deepwell Storage Plate

2.2 mL deepwell storage plates for the culture or storage of large samples.

Brand:  Thermo Scientific™ AB-0661/W

Code : 03

Additional Details : Weight : 4.70000kg

 View more versions of this product

Product Code. 10622664

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Highlights
  • Designed for optimal sample recovery
  • Each square well is specifically designed with an independent sealing rim to prevent cross-contamination
  • U-bottom (AB-0661) to aid resuspension or conical bottom (AB-0932) for ease of sample retrieval
Specifications

Specifications

Square-Well
White
50
Square well
For Research Use Only. Not for use in diagnostic procedures.
U shaped
Feature square well geometry which promotes efficient sample aeration, and U-bottomed wells to aid in re-suspension
2.2mL
Individual
Polypropylene Storage Plate
Videos
Documentation

Technical Documents