missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Methylphenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62512.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477506

  • £99.64 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Methylphenyl)thiazolidine-4-carboxylic acid
223.29
MFCD00112828
2-(o-Tolyl)thiazolidine-4-carboxylic acid

Specifications

2-(2-Methylphenyl)thiazolidine-4-carboxylic acid
Solid
223.29
2-(o-Tolyl)thiazolidine-4-carboxylic acid
1g
97%
MFCD00112828
Videos
SDS