missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1H-imidazol-1-yl)benzonitrile, 97%, Thermo Scientific™

Catalog Number p-3527234
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
892502-27-1
C11H9N3
183.214
MFCD09065013
NCQQCFZREDABTC-UHFFFAOYSA-N
2-2-methyl-1h-imidazol-1-yl benzonitrile, 2-2-methylimidazol-1-yl benzonitrile, 2-2-methylimidazolyl benzenecarbonitrile, benzonitrile,2-2-methyl-1h-imidazol-1-yl
24229637
2-(2-methylimidazol-1-yl)benzonitrile
CC1=NC=CN1C2=CC=CC=C2C#N
This item is not returnable. View return policy
892502-27-1
C11H9N3
183.214
MFCD09065013
NCQQCFZREDABTC-UHFFFAOYSA-N
2-2-methyl-1h-imidazol-1-yl benzonitrile, 2-2-methylimidazol-1-yl benzonitrile, 2-2-methylimidazolyl benzenecarbonitrile, benzonitrile,2-2-methyl-1h-imidazol-1-yl
24229637
2-(2-methylimidazol-1-yl)benzonitrile
CC1=NC=CN1C2=CC=CC=C2C#N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-1H-imidazol-1-yl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'