missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1,3-thiazol-4-yl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-3586620
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
65032-66-8
C11H9NO2S
219.258
MFCD09879980
BNRSCIXYHUTATP-UHFFFAOYSA-N
2-2-methyl-1,3-thiazol-4-yl benzoic acid, benzoic acid,2-2-methyl-4-thiazolyl, 2-2-methylthiazol-4-yl benzoic acid, 2-2-methyl-thiazol-4-yl-benzoic acid, 4-2-carboxyphenyl-2-methyl-1,3-thiazole
12384552
2-(2-methyl-1,3-thiazol-4-yl)benzoic acid
CC1=NC(=CS1)C2=CC=CC=C2C(=O)O
This item is not returnable. View return policy
65032-66-8
C11H9NO2S
219.258
MFCD09879980
BNRSCIXYHUTATP-UHFFFAOYSA-N
2-2-methyl-1,3-thiazol-4-yl benzoic acid, benzoic acid,2-2-methyl-4-thiazolyl, 2-2-methylthiazol-4-yl benzoic acid, 2-2-methyl-thiazol-4-yl-benzoic acid, 4-2-carboxyphenyl-2-methyl-1,3-thiazole
12384552
2-(2-methyl-1,3-thiazol-4-yl)benzoic acid
CC1=NC(=CS1)C2=CC=CC=C2C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'