missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Methoxyphenyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62646.03

Code : NEW

Additional Details : CAS Number : 40790-78-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459148

  • £91.90 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Methoxyphenyl)thiazolidine
195.28
2-(2-Thiazolidinyl)anisole
40790-78-1
MFCD00805017

Specifications

2-(2-Methoxyphenyl)thiazolidine
Liquid
195.28
1g
40790-78-1
97%
MFCD00805017
2-(2-Thiazolidinyl)anisole
Videos
SDS