missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Methoxyphenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62699.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487746

  • £128.26 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Methoxyphenyl)thiazolidine-4-carboxylic acid
239.29
MFCD01821271

Specifications

2-(2-Methoxyphenyl)thiazolidine-4-carboxylic acid
97%
MFCD01821271
Solid
239.29
1g
Videos
SDS