missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methoxyphenyl)benzimidazole, 95%, Thermo Scientific™

Catalog Number 15481419
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
6528-85-4
C14H12N2O
224.263
MFCD00447053
JNFBDHNAZPBWTF-UHFFFAOYSA-N
2-2-methoxyphenyl-1h-benzimidazole, 1h-benzimidazole, 2-2-methoxyphenyl, 2-2-methoxyphenyl-1h-benzo d imidazole, 2-2-methoxyphenyl-1h-1,3-benzodiazole, 2-2-methoxy-phenyl-1h-benzoimidazole, 1h-benzo d imidazole,2-2-methoxyphenyl
759420
2-(2-methoxyphenyl)-1H-benzimidazole
COC1=CC=CC=C1C2=NC3=CC=CC=C3N2
This item is not returnable. View return policy
6528-85-4
C14H12N2O
224.263
MFCD00447053
JNFBDHNAZPBWTF-UHFFFAOYSA-N
2-2-methoxyphenyl-1h-benzimidazole, 1h-benzimidazole, 2-2-methoxyphenyl, 2-2-methoxyphenyl-1h-benzo d imidazole, 2-2-methoxyphenyl-1h-1,3-benzodiazole, 2-2-methoxy-phenyl-1h-benzoimidazole, 1h-benzo d imidazole,2-2-methoxyphenyl
759420
2-(2-methoxyphenyl)-1H-benzimidazole
COC1=CC=CC=C1C2=NC3=CC=CC=C3N2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 224.26
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(2-Methoxyphenyl)benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'