missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Methoxyphenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64294.MD

Code : NEW

Additional Details : CAS Number : 6528-85-4 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15401429

  • £32.85 / 250mg

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

6528-85-4
224.263
JNFBDHNAZPBWTF-UHFFFAOYSA-N
759420
COC1=CC=CC=C1C2=NC3=CC=CC=C3N2
C14H12N2O
MFCD00447053
2-2-methoxyphenyl-1h-benzimidazole, 1h-benzimidazole, 2-2-methoxyphenyl, 2-2-methoxyphenyl-1h-benzo d imidazole, 2-2-methoxyphenyl-1h-1,3-benzodiazole, 2-2-methoxy-phenyl-1h-benzoimidazole, 1h-benzo d imidazole,2-2-methoxyphenyl
2-(2-methoxyphenyl)-1H-benzimidazole

Specifications

2-(2-Methoxyphenyl)benzimidazole
6528-85-4
MFCD00447053
JNFBDHNAZPBWTF-UHFFFAOYSA-N
2-(2-methoxyphenyl)-1H-benzimidazole
759420
Solid
250mg
C14H12N2O
2-2-methoxyphenyl-1h-benzimidazole, 1h-benzimidazole, 2-2-methoxyphenyl, 2-2-methoxyphenyl-1h-benzo d imidazole, 2-2-methoxyphenyl-1h-1,3-benzodiazole, 2-2-methoxy-phenyl-1h-benzoimidazole, 1h-benzo d imidazole,2-2-methoxyphenyl
COC1=CC=CC=C1C2=NC3=CC=CC=C3N2
224.263
224.26
95%
Videos
SDS Documents
Certificates