missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Methoxyphenyl)-5-methylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64063.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15461089

  • £91.90 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Methoxyphenyl)-5-methylbenzimidazole
238.29
MFCD11849219

Specifications

2-(2-Methoxyphenyl)-5-methylbenzimidazole
95%
MFCD11849219
Solid
238.29
1g
Videos
SDS