missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyethyl)pyridine, 99%, Thermo Scientific™

Catalog Number p-144854
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
103-74-2
MFCD00006364
BXGYBSJAZFGIPX-UHFFFAOYSA-N
2-pyridineethanol, 2-2-hydroxyethyl pyridine, 2-2-pyridyl ethanol, pyridine-2-ethanol, 2-pyridin-2-yl ethan-1-ol, 2-pyridin-2-yl ethanol, 2-pyridinethanol, 2-pyridylethanol, 2-pyridine ethanol, 2-beta-hydroxyethyl pyridine
7675
2-pyridin-2-ylethanol
C1=CC=NC(=C1)CCO
This item is not returnable. View return policy
103-74-2
MFCD00006364
BXGYBSJAZFGIPX-UHFFFAOYSA-N
2-pyridineethanol, 2-2-hydroxyethyl pyridine, 2-2-pyridyl ethanol, pyridine-2-ethanol, 2-pyridin-2-yl ethan-1-ol, 2-pyridin-2-yl ethanol, 2-pyridinethanol, 2-pyridylethanol, 2-pyridine ethanol, 2-beta-hydroxyethyl pyridine
7675
2-pyridin-2-ylethanol
C1=CC=NC(=C1)CCO

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.0900g/mL
Boiling Point 235°C to 238°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H9NO
Linear Formula (C5H4N)CH2CH2OH
Refractive Index 1.5350 to 1.537
Quantity 5 g
Beilstein 21, 50
Fieser 12,416
Specific Gravity 1.09
Solubility Information Solubility in water: freely soluble
Molecular Weight (g/mol) 123.15
Viscosity 40 mPa.s (20°C)
Formula Weight 123.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(2-Hydroxyethyl)pyridine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Hydroxyethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'