missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Furyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62080.03

Code : NEW

Additional Details : CAS Number : 115299-13-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15438668

  • £119.78 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Furyl)thiazolidine-4-carboxylic acid
115299-13-3
199.23
MFCD00170602

Specifications

2-(2-Furyl)thiazolidine-4-carboxylic acid
Solid
199.23
1g
115299-13-3
97%
MFCD00170602
Videos
SDS