missing translation for 'onlineSavingsMsg'
Learn More

[2-(2-Furyl)phenyl]methanol, ≥97%, Thermo Scientific™

Catalog Number 10664215
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
139697-88-4
C11H10O2
174.20
MFCD06203101
NBLMKRIYULDNBF-UHFFFAOYSA-N
2-2-furyl phenyl methanol, 2-furan-2-yl phenyl methanol, 2-fur-2-yl benzyl alcohol, 2-2-furyl benzenemethanol, 2-fur-2-yl phenyl methanol, benzenemethanol,2-2-furanyl, 2-2-furyl phenyl methan-1-ol
10855985
[2-(furan-2-yl)phenyl]methanol
OCC1=CC=CC=C1C1=CC=CO1
This item is not returnable. View return policy
139697-88-4
C11H10O2
174.20
MFCD06203101
NBLMKRIYULDNBF-UHFFFAOYSA-N
2-2-furyl phenyl methanol, 2-furan-2-yl phenyl methanol, 2-fur-2-yl benzyl alcohol, 2-2-furyl benzenemethanol, 2-fur-2-yl phenyl methanol, benzenemethanol,2-2-furanyl, 2-2-furyl phenyl methan-1-ol
10855985
[2-(furan-2-yl)phenyl]methanol
OCC1=CC=CC=C1C1=CC=CO1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material [2-(2-Furyl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'