Learn More
Thermo Scientific Maybridge 2-(2-Furyl)benzaldehyde, 1g, 97%
Brand: Thermo Scientific Maybridge CC46204DA
2-(2-Furyl)benzaldehyde, 1g, 97%
Code : 31
Additional Details : CAS Number : 16191-32-5 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC46204DA
2-(2-Furyl)benzaldehyde, 1g, 97%
Code : 31
Additional Details : CAS Number : 16191-32-5 Weight : 0.05100kg