missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Furyl)benzaldehyde, ≥97%, Maybridge

Brand:  Maybridge CC46204EA

Code : 31

Additional Details : CAS Number : 16191-32-5 Weight : 0.06000kg

Quantity 10g

 View more versions of this product

Product Code. 10255574

  • £269.00 / 10g


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Furyl)benzaldehyde
96.5
97%
C11H8O2
MFCD03990475
16191-32-5
100.0
97%
172.18

Specifications

2-(2-Furyl)benzaldehyde
96.5
97%
C11H8O2
MFCD03990475
16191-32-5
100.0
97%
172.18
10g
Videos
SDS