missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Fluorophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64457.06

Code : NEW

Additional Details : CAS Number : 321-51-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15471669

  • £543.78 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-(2-Fluorophenyl)benzimidazole
321-51-7
212.23
MFCD01086283

Specifications

2-(2-Fluorophenyl)benzimidazole
321-51-7
95%
MFCD01086283
5g
Solid
212.23
Videos
SDS