missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Diphenylglycine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.
Solubility
It is sparingly soluble in water.
Notes
Incompatible with oxidizing agents.
2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.
Solubility
It is sparingly soluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | ∼246°C (decomposition) |
Solubility Information | It is sparingly soluble in water. |
Formula Weight | 227.26 |
Percent Purity | 98% |
Quantity | 5 g |
Chemical Name or Material | 2,2-Diphenylglycine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'