missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylethylamine, 96%, Thermo Scientific Chemicals

Catalog Number 10469600
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3963-62-0
C14H15N
197.28
MFCD00008143
RXMTUVIKZRXSSM-UHFFFAOYSA-N
2,2-diphenylethylamine, 2,2-diphenyl-ethylamine, 1-amino-2,2-diphenylethane, 2,2-diphenylethan-1-amine, chembl10780, pubchem7986, acmc-1cswz, 2,2-diphenylethyl-amine, 2,2-di phenyl ethanamine
77575
2,2-diphenylethanamine
NCC(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
3963-62-0
C14H15N
197.28
MFCD00008143
RXMTUVIKZRXSSM-UHFFFAOYSA-N
2,2-diphenylethylamine, 2,2-diphenyl-ethylamine, 1-amino-2,2-diphenylethane, 2,2-diphenylethan-1-amine, chembl10780, pubchem7986, acmc-1cswz, 2,2-diphenylethyl-amine, 2,2-di phenyl ethanamine
77575
2,2-diphenylethanamine
NCC(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 43°C to 49°C
Color White to Yellow
Boiling Point 139°C to 140°C (1.2 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (C6H5)2CHCH2NH2
Quantity 25 g
Beilstein 12, 1327
Formula Weight 197.28
Percent Purity 96%
Physical Form Fused Solid
Chemical Name or Material 2, 2-Diphenylethylamine, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 2,2-Diphenylethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'