missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylethanol, 97%, Thermo Scientific Chemicals

Catalog Number p-3757774
23.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1883-32-5
C14H14O
198.26
MFCD00004729
NYLOEXLAXYHOHH-UHFFFAOYSA-N
beta-phenylphenethyl alcohol, 2,2-diphenyl ethanol, benzeneethanol, .beta.-phenyl, acmc-20aojg, 2,2-diphenyl-ethanol, 2,2-diphenyl-1-ethanol, 2,2-diphenylethan-1-ol, benzeneethanol, b-phenyl, 2,2-diphenylethanol, ksc539o6l
74662
2,2-diphenylethanol
C1=CC=C(C=C1)C(CO)C2=CC=CC=C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1883-32-5
C14H14O
198.26
MFCD00004729
NYLOEXLAXYHOHH-UHFFFAOYSA-N
beta-phenylphenethyl alcohol, 2,2-diphenyl ethanol, benzeneethanol, .beta.-phenyl, acmc-20aojg, 2,2-diphenyl-ethanol, 2,2-diphenyl-1-ethanol, 2,2-diphenylethan-1-ol, benzeneethanol, b-phenyl, 2,2-diphenylethanol, ksc539o6l
74662
2,2-diphenylethanol
C1=CC=C(C=C1)C(CO)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 53°C to 56°C
Color White
Boiling Point 160°C to 162°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CHCH2OH
Quantity 2 g
Beilstein 06, II, 640
Formula Weight 198.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2-Diphenylethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2,2-Diphenylethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'