missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,2-Diphenyl-N-propylacetamide, 95%

Brand:  Alfa Aesar™ H65924.03

Code : NEW

Additional Details : CAS Number : 20461-07-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402669

  • £128.26 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

2,2-Diphenyl-N-propylacetamide
253.35
2119043
20461-07-8
MFCD00430992

Specifications

2,2-Diphenyl-N-propylacetamide
20461-07-8
95%
MFCD00430992
1g
Solid
253.35
2119043
Videos
SDS