missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,2-Diphenyl-N-[4-(trifluoromethyl)phenyl]acetamide, 95%

Brand:  Alfa Aesar™ H65989.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15430077

  • £74.73 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

2,2-Diphenyl-N-[4-(trifluoromethyl)phenyl]acetamide
355.36
MFCD01489467
N-[4-(Trifluoromethyl)phenyl]-2,2-diphenylacetamide

Specifications

2,2-Diphenyl-N-[4-(trifluoromethyl)phenyl]acetamide
Solid
355.36
N-[4-(Trifluoromethyl)phenyl]-2,2-diphenylacetamide
1g
95%
MFCD01489467
Videos
SDS