missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2,2-Diphenyl-N-[3-(trifluoromethyl)phenyl]acetamide, 95%
Brand: Alfa Aesar™ H65945.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2,2-Diphenyl-N-[3-(trifluoromethyl)phenyl]acetamide | |
355.36 | |
MFCD00370058 | |
N-[3-(Trifluoromethyl)phenyl]-2,2-diphenylacetamide |
Specifications
2,2-Diphenyl-N-[3-(trifluoromethyl)phenyl]acetamide | |
Solid | |
355.36 | |
N-[3-(Trifluoromethyl)phenyl]-2,2-diphenylacetamide |
1g | |
95% | |
MFCD00370058 |