missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylbutane 98%, ACROS Organics™

Brand:  Acros Organics 116090025

Code : 29

Additional Details : CAS Number : 75-83-2 Weight : 3.12000kg Transport : UN number : 1208 Chem class : 1 Pack group : II

Quantity 2.5L
Packaging Glass bottle

 View more versions of this product

Product Code. 10297283

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2, 2-Dimethylbutane
75-83-2
100.0
98%
C2H5C(CH3)3
MFCD00009321
98%
97.5
≥97.5%
C6H14
86.18
Neohexane

Specifications

2, 2-Dimethylbutane
Authentic
2.5L
75-83-2
100.0
1.3675 to 1.3695 (20°C, 589nm)
≥97.5%
C6H14
86.18
01, 150
0.6490g/mL
Clear colorless to slightly yellow
98%
97.5
Glass bottle
Liquid
98%
C2H5C(CH3)3
MFCD00009321
Neohexane
Videos
SDS