missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethyl-6-chromanesulfonyl chloride, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 260.74 |
Percent Purity | 90% |
Chemical Name or Material | 2, 2-Dimethyl-6-chromanesulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'