missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-3-butenoic acid, 95%, Thermo Scientific Chemicals

Catalog Number 15461099
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10276-09-2
C6H10O2
114.144
MFCD00196967
SCFWAOWWAANBPY-UHFFFAOYSA-N
139122
2,2-dimethylbut-3-enoic acid
CC(C)(C=C)C(=O)O
This item is not returnable. View return policy
10276-09-2
C6H10O2
114.144
MFCD00196967
SCFWAOWWAANBPY-UHFFFAOYSA-N
139122
2,2-dimethylbut-3-enoic acid
CC(C)(C=C)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -6°C
Color Colorless
Density 0.957 g/mL
Boiling Point 185°C
Refractive Index 1.4295
Quantity 1 g
UN Number UN3265
Beilstein 1700758
Synonym 3-butenoic acid, 2,2-dimethyl, 2,2-dimethyl-but-3-enoic acid, 2,2-dimethyl-3-butenoic acid, ch2=chc ch3 2cooh, 3-butenoic acid,2,2-dimethyl
Formula Weight 114.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2,2-Dimethyl-3-butenoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'