missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethyl-3-butenoic acid, 95%, Thermo Scientific Chemicals
324.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -6°C |
Color | Colorless |
Density | 0.957 g/mL |
Boiling Point | 185°C |
Refractive Index | 1.4295 |
Quantity | 5 g |
UN Number | UN3265 |
Beilstein | 1700758 |
Synonym | 3-butenoic acid, 2,2-dimethyl, 2,2-dimethyl-but-3-enoic acid, 2,2-dimethyl-3-butenoic acid, ch2=chc ch3 2cooh, 3-butenoic acid,2,2-dimethyl |
Formula Weight | 114.14 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'