missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%

£52.70 - £209.00

Chemical Identifiers

CAS 866089-28-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD11603419
InChI Key WILBVUMFTFLAHJ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazine, 2,2-dimethyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-2,2-dimethyl, 2,2-dimethyl-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-2,2-dimethyl
PubChem CID 23587101
IUPAC Name 2,2-dimethyl-3,4-dihydro-1,4-benzoxazine
SMILES CC1(CNC2=CC=CC=C2O1)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15406236
SDS View Documents
Alfa Aesar™
H61058.MD
250mg
£52.70
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 22-10-2021
Please to purchase this item. Need a web account? Register with us today!
15497548
SDS View Documents
Alfa Aesar™
H61058.03
1g
£209.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 22-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications
It possess various biological activities.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications

Chemical Identifiers

866089-28-3
163.22
WILBVUMFTFLAHJ-UHFFFAOYSA-N
23587101
CC1(CNC2=CC=CC=C2O1)C
C10H13NO
MFCD11603419
2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazine, 2,2-dimethyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-2,2-dimethyl, 2,2-dimethyl-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-2,2-dimethyl
2,2-dimethyl-3,4-dihydro-1,4-benzoxazine

Specifications

2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine
866089-28-3
MFCD11603419
WILBVUMFTFLAHJ-UHFFFAOYSA-N
2,2-dimethyl-3,4-dihydro-1,4-benzoxazine
23587101
97%
250mg
C10H13NO
2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazine, 2,2-dimethyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-2,2-dimethyl, 2,2-dimethyl-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-2,2-dimethyl
CC1(CNC2=CC=CC=C2O1)C
163.22
163.22
SDS
Product Certifications