missing translation for 'onlineSavingsMsg'
Learn More

(2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-yl)methylamine, ≥97%, Thermo Scientific™

Catalog Number p-3175066
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
868755-44-6
C11H15NO
177.25
MFCD08271885
BQEPNISHAAOSFF-UHFFFAOYSA-N
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methylamine, 2,2-dimethyl-2,3-dihydrobenzofuran-7-yl methanamine, 2,2-dimethyl-3h-1-benzofuran-7-yl methanamine, 1-2,2-dimethyl-3h-1-benzofuran-7-yl methanamine, 7-benzofuranmethanamine,2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methanamine, 2,3-dihydro-2,2-dimethyl-1-benzofuran-7-yl methylamine, 2,2-dimethyl-2,3-dihydrobenzo 2,3-b furan-7-yl methylamine, 1-2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methanamine, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-yl methylamine
18525713
CC1(C)CC2=C(O1)C(CN)=CC=C2
This item is not returnable. View return policy
868755-44-6
C11H15NO
177.25
MFCD08271885
BQEPNISHAAOSFF-UHFFFAOYSA-N
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methylamine, 2,2-dimethyl-2,3-dihydrobenzofuran-7-yl methanamine, 2,2-dimethyl-3h-1-benzofuran-7-yl methanamine, 1-2,2-dimethyl-3h-1-benzofuran-7-yl methanamine, 7-benzofuranmethanamine,2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methanamine, 2,3-dihydro-2,2-dimethyl-1-benzofuran-7-yl methylamine, 2,2-dimethyl-2,3-dihydrobenzo 2,3-b furan-7-yl methylamine, 1-2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl methanamine, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-yl methylamine
18525713
CC1(C)CC2=C(O1)C(CN)=CC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name (2,2-dimethyl-3H-1-benzofuran-7-yl)methanamine
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material (2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'