2,2'-Dimethoxy-1,1'-binaphthyl, 98%, ACROS Organics™


Overview
£89.68 valid until 31/03/2017
Log in to get the promotional price



Brand: Acros Organics

Manufacturer Part Number: 342360050

5GR 2,2'-Dimethoxy-1,1'-binaphthyl, 98%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 2960-93-2 Weight: 0.05500kg


Packaging Glass bottle
Quantity 5g

Product Code. 10613332

Quantity Price
1 £ 224.2 / 5g
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2, 2'-Dimethoxy-1, 1'-binaphthyl
Name Note 98%
CAS 2960-93-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C22H18O2
Formula Weight 314.38
MDL Number MFCD00091146
Synonym 1, 1'-Binaphthol dimethyl ether

Specification

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 314.38
Quantity 5g
Color White to beige
CAS Min % 97.5
CAS 2960-93-2
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00091146
Packaging Glass bottle
Name Note 98%
Molecular Formula C22H18O2
CAS Max % 100.0
Chemical Name or Material 2, 2'-Dimethoxy-1, 1'-binaphthyl
TSCA TSCA
Synonym 1, 1'-Binaphthol dimethyl ether
Melting Point 198° to 202°C
Documentation

Technical documents