missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoroethanol, 97%, Thermo Scientific Chemicals

Catalog Number p-7026277
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
359-13-7
C2H4F2O
82.05
MFCD00042248
VOGSDFLJZPNWHY-UHFFFAOYSA-N
2,2-difluoroethan-1-ol, 2,2-difluoro-ethanol, ethanol, 2,2-difluoro, chf2ch2oh, 2,2-difluroethanol, 2,2-difluorethanol, 2,2-difluoro ethanol, 2,2-difluoroethylalcohol, difluoroethanol related, acmc-209ik0
136187
2,2-difluoroethanol
C(C(F)F)O
This item is not returnable. View return policy
359-13-7
C2H4F2O
82.05
MFCD00042248
VOGSDFLJZPNWHY-UHFFFAOYSA-N
2,2-difluoroethan-1-ol, 2,2-difluoro-ethanol, ethanol, 2,2-difluoro, chf2ch2oh, 2,2-difluroethanol, 2,2-difluorethanol, 2,2-difluoro ethanol, 2,2-difluoroethylalcohol, difluoroethanol related, acmc-209ik0
136187
2,2-difluoroethanol
C(C(F)F)O

2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

Solubility
soluble in chloroform, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY
Melting Point -28°C
Density 1.296
Boiling Point 95°C to 96°C
Flash Point 3°C (37°F)
Refractive Index 1.3335
Quantity 1 g
UN Number UN1987
Beilstein 1731617
Solubility Information soluble in chloroform,ethanol.
Formula Weight 82.05
Percent Purity 97%
Chemical Name or Material 2,2-Difluoroethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'