missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diethoxyacetophenone, 98%, ACROS Organics™

Brand:  Acros Organics 210621000

Code : 29

Additional Details : CAS Number : 6175-45-7 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10592991

  • £78.90 / 100g


Estimated Shipment
17-05-2018
Add to basket

Specifications

Chemical Identifiers

2, 2-Diethoxyacetophenone
6175-45-7
100.0
96%
C6H5COCH(OC2H5)2
MFCD00009659
98%
97.5
≥97.5%
C12H16O3
208.26
2, 2-Diethoxy-1-phenylethanone, DEAP

Specifications

2, 2-Diethoxyacetophenone
98%
97.5
1.0300g/mL
Glass bottle
Liquid
96%
C6H5COCH(OC2H5)2
MFCD00009659
07, I, 361
100g
6175-45-7
100.0
Authentic
1.4985 to 1.5005 (20°C, 589nm)
≥97.5%
C12H16O3
208.26
Clear yellow to salmon
2, 2-Diethoxy-1-phenylethanone, DEAP
Videos
SDS