Alfa Aesar™ 2-(2-Chlorophenyl)-5-methylbenzimidazole, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64997.03

1GR 2-(2-Chlorophenyl)-5-methylbenzimidazole, 95% 1g

UNSPSC: 12352005

Code: NEW

Additional Details:
Additional Details: CAS Number: 14225-76-4 Weight: 0.00100kg


Quantity 1g

Product Code. 15432439

Quantity Price
1 £ 91.9 / 1g
Estimated Shipment date
from Supplier 01-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-(2-Chlorophenyl)-5-methylbenzimidazole
CAS 14225-76-4
Formula Weight 242.71
MDL Number MFCD01836513

Specification

Formula Weight 242.71
Chemical Name or Material 2-(2-Chlorophenyl)-5-methylbenzimidazole
Physical Form Solid
Assay 95%
MDL Number MFCD01836513
CAS 14225-76-4
Quantity 1g