missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Chloroethoxy)ethanol, 99%, Thermo Scientific Chemicals
50.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.
Solubility
Miscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.
Solubility
Miscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Density | 1.18 |
Boiling Point | 185°C |
Flash Point | 90°C (194°F) |
Linear Formula | ClCH2CH2OCH2CH2OH |
Refractive Index | 1.4535 |
Quantity | 250 g |
Beilstein | 506015 |
Solubility Information | Miscible with water. |
Formula Weight | 124.57 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'