missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Chloro-6-fluorobenzyloxy)benzaldehyde, 96%

Brand:  Alfa Aesar™ 044458.03

Code : NEW

Additional Details : CAS Number : 336880-01-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11420780

  • £191.01 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

2- (2-Chloro-6-fluorobenzyloxy)benzaldehyde
C14H10ClFO2
MFCD01590675
336880-01-4
264.7

Specifications

2- (2-Chloro-6-fluorobenzyloxy)benzaldehyde
1g
Powder
C14H10ClFO2
MFCD01590675
Air & Moisture Sensitive
336880-01-4
96%
264.7
Videos
SDS