missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromoethyl)-1,3-dioxolane, 96%, Thermo Scientific Chemicals

Catalog Number p-3559297
Click to view available options
Quantity:
10 g
25 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18742-02-4
C5H9BrO2
181.03
MFCD00003216
GGZQLTVZPOGLCC-UHFFFAOYSA-N
2-2-bromoethyl-1,3-dioxolane, 1,3-dioxolane, 2-2-bromoethyl, bromoethyl-1,3-dioxolane, acmc-1bol0, 2-2-bromoethyl-dioxolane, ksc496m8b, 2-bromoethyl-1,3-dioxolane, 2 2-bromoethyl-1,3-dioxolane, 2-2-bromoethyl-1,3-dioxolan
87776
2-(2-bromoethyl)-1,3-dioxolane
BrCCC1OCCO1
This item is not returnable. View return policy
18742-02-4
C5H9BrO2
181.03
MFCD00003216
GGZQLTVZPOGLCC-UHFFFAOYSA-N
2-2-bromoethyl-1,3-dioxolane, 1,3-dioxolane, 2-2-bromoethyl, bromoethyl-1,3-dioxolane, acmc-1bol0, 2-2-bromoethyl-dioxolane, ksc496m8b, 2-bromoethyl-1,3-dioxolane, 2 2-bromoethyl-1,3-dioxolane, 2-2-bromoethyl-1,3-dioxolan
87776
2-(2-bromoethyl)-1,3-dioxolane
BrCCC1OCCO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Density 1.5400g/mL
Boiling Point 68°C to 70°C (8.0 mmHg)
Flash Point 65°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4783 to 1.4803
Quantity 10 g
Fieser 07,37; 11,78
Specific Gravity 1.54
Solubility Information Solubility in water: immiscible
Formula Weight 181.03
Percent Purity 96%
Chemical Name or Material 2-(2-Bromoethyl)-1, 3-dioxolane
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Bromoethyl)-1,3-dioxolane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'