missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bithiophene-5-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog Number 15497847
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
479719-88-5
C14H17BO2S2
292.22
MFCD04039974
HPOQARMSOPOZMW-UHFFFAOYSA-N
2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol ester
3592790
4,4,5,5-tetramethyl-2-(5-thiophen-2-ylthiophen-2-yl)-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1
This item is not returnable. View return policy
479719-88-5
C14H17BO2S2
292.22
MFCD04039974
HPOQARMSOPOZMW-UHFFFAOYSA-N
2,2'-bithiophene-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,2'-bithiophene, 2-2,2'-bithiophen-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-5-thiophen-2-yl thiophen-2-yl-1,3,2-dioxaborolane, acmc-209kb7, 4,4,5,5-tetramethyl-2-5-thiophen-2-ylthiophen-2-yl-1,3,2-dioxaborolane, 2,2'-bithiophene-5'-boronic acid pinacol ester
3592790
4,4,5,5-tetramethyl-2-(5-thiophen-2-ylthiophen-2-yl)-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 35°C to 38°C
Color Blue-Green to White
Quantity 5 g
Formula Weight 292.2
Percent Purity 98%
Physical Form Fused Solid
Chemical Name or Material 2,2'-Bithiophene-5-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'