missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bipyridine, ACS, 98%, Thermo Scientific Chemicals

Catalog Number 11317318
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
366-18-7
C10H8N2
156.19
MFCD00006212
ROFVEXUMMXZLPA-UHFFFAOYSA-N
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
1474
CHEBI:30351
C1=CC=C(N=C1)C1=CC=CC=N1
This item is not returnable. View return policy
366-18-7
C10H8N2
156.19
MFCD00006212
ROFVEXUMMXZLPA-UHFFFAOYSA-N
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
1474
CHEBI:30351
C1=CC=C(N=C1)C1=CC=CC=N1

2,2'-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70°C
Color White
Boiling Point 272°C to 273°C
Flash Point 121°C (249°F)
Quantity 10 g
UN Number UN2811
Beilstein 113089
Merck Index 14,3347
Solubility Information Soluble in organic solvents and slightly soluble in water.
IUPAC Name 2-pyridin-2-ylpyridine
Formula Weight 156.19
Percent Purity 98%
Grade ACS
Physical Form Crystalline
Chemical Name or Material 2,2'-Bipyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'