missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Biphenol, 99%, Thermo Scientific Chemicals

Catalog Number p-66905
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
1806-29-7
C12H10O2
186.21
MFCD00002210
IMHDGJOMLMDPJN-UHFFFAOYSA-N
2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl
15731
CHEBI:28970
2-(2-hydroxyphenyl)phenol
OC1=CC=CC=C1C1=CC=CC=C1O
This item is not returnable. View return policy
1806-29-7
C12H10O2
186.21
MFCD00002210
IMHDGJOMLMDPJN-UHFFFAOYSA-N
2,2'-biphenol, 1,1'-biphenyl-2,2'-diol, 2,2'-dihydroxybiphenyl, biphenyl-2,2'-diol, o,o'-diphenol, o,o'-biphenol, o-dihydroxydiphenyl, o,o'-dihydroxybiphenyl, 2,2'-biphenyldiol, 2,2'-dihydroxydiphenyl
15731
CHEBI:28970
2-(2-hydroxyphenyl)phenol
OC1=CC=CC=C1C1=CC=CC=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 108°C to 111°C
Color Beige to Beige-Gray
Density 0.8300g/mL
Boiling Point 315°C
Flash Point 154°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H4C6H4OH
Quantity 100 g
Beilstein 06, 989
Specific Gravity 0.83
Solubility Information Solubility in water: insoluble
Formula Weight 186.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2,2′-Biphenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,2'-Biphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'