missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Aminoethyl)thio]ethan-1-ol, ≥90%, Maybridge

Brand:  Maybridge SB01634EA

Code : 31

Additional Details : CAS Number : 24304-84-5 Weight : 0.06000kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 10744393

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-[(2-Aminoethyl)thio]ethan-1-ol
90.0
90%
NH2CH2CH2SCH2CH2OH
24304-84-5
90%
C4H11NOS
121.2

Specifications

2-[(2-Aminoethyl)thio]ethan-1-ol
90.0
90%
90%
NH2CH2CH2SCH2CH2OH
24304-84-5
1.1200g/mL
10g
C4H11NOS
121.2
Videos
SDS