missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%, ACROS Organics™

Brand:  Acros Organics 301450010

Code : 29

Additional Details : CAS Number : 15871-57-5 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10368803

  • £206.10 / 1g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid
15871-57-5
100.0
96%
215.27
4-Amino-4-carboxy-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-Amino-4-carboxy-2, 2, 6, TOAC
96%
95.0
≥95% (GC)
C10H19N2O3
MFCD00800575

Specifications

2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid
1g
15871-57-5
100.0
Glass bottle
≥95% (GC)
C10H19N2O3
MFCD00800575
White to off-white
96%
95.0
Authentic
Crystalline Powder
96%
215.27
4-Amino-4-carboxy-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-Amino-4-carboxy-2, 2, 6, TOAC
Videos
SDS