missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethyl-4-piperidinol, 99%, Thermo Scientific Chemicals

Catalog Number p-198795
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2403-88-5
C9H19NO
157.26
MFCD00005983
VDVUCLWJZJHFAV-UHFFFAOYSA-N
2,2,6,6-tetramethyl-4-piperidinol, lastar a, 4-piperidinol, 2,2,6,6-tetramethyl, 4-hydroxy-2,2,6,6-tetramethylpiperidine, 2,2,6,6-tetramethyl-4-hydroxypiperidine, unii-is23q8egz6, is23q8egz6, 2,2,6,6-tetramethyl-piperidin-4-ol, pubchem23863
75471
2,2,6,6-tetramethylpiperidin-4-ol
CC1(CC(CC(N1)(C)C)O)C
This item is not returnable. View return policy
2403-88-5
C9H19NO
157.26
MFCD00005983
VDVUCLWJZJHFAV-UHFFFAOYSA-N
2,2,6,6-tetramethyl-4-piperidinol, lastar a, 4-piperidinol, 2,2,6,6-tetramethyl, 4-hydroxy-2,2,6,6-tetramethylpiperidine, 2,2,6,6-tetramethyl-4-hydroxypiperidine, unii-is23q8egz6, is23q8egz6, 2,2,6,6-tetramethyl-piperidin-4-ol, pubchem23863
75471
2,2,6,6-tetramethylpiperidin-4-ol
CC1(CC(CC(N1)(C)C)O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 129°C to 133°C
Color Beige or White
Boiling Point 212°C to 215°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 12
Solubility Information Solubility in water: 130g/l (23 c)
Formula Weight 157.26
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2,2,6,6-Tetramethyl-4-piperidinol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,2,6,6-Tetramethyl-4-piperidinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'