missing translation for 'onlineSavingsMsg'
Learn More

(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium, 98%, Thermo Scientific Chemicals

Catalog Number p-7050552
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
22441-13-0
MFCD00142754
Lithium 2,2,6,6-tetramethyl-3,5-heptanedionate; Li(TMHD)
This item is not returnable. View return policy
22441-13-0
MFCD00142754
Lithium 2,2,6,6-tetramethyl-3,5-heptanedionate; Li(TMHD)

Used as reactant for synthesis of Bis-cycloiridiated complexes for electroluminescent studies, cationic imido alkylidene complexes, polyaromatic ligands containing pyrene and mono-cyclometalated platinum complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 127°C
Boiling Point 180°C (0.05 mmHg)
Quantity 1 g
Solubility Information Soluble in acetone and methanol.
Formula Weight 190.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'