missing translation for 'onlineSavingsMsg'
Learn More

2,2':6',2''-Terpyridine-4'-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog Number 15457817
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
148332-36-9
C16H11N3O2
277.28
MFCD04114290
ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
2,2':6',2-terpyridine-4'-carboxylic acid, 2,2':6',2-terpyridine-4'-carboxylicacid, 2,6-bis pyridin-2-yl pyridine-4-carboxylic acid, 6-pyridin-2-yl-2,2'-bipyridine-4-carboxylic acid, acmc-1c3v7, 4'-carboxy-2,2':6',2-terpyridine, 2,2:6,2'-terpyridine-4-carboxylic acid, 1∼2∼,2∼2∼:2∼6∼,3∼2∼-terpyridine-2∼4∼-carboxylic acid, 2,2\\':6\\',2\\'\\'-terpyridine-4\\'-carboxylic acid
2762749
2,6-dipyridin-2-ylpyridine-4-carboxylic acid
OC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
This item is not returnable. View return policy
148332-36-9
C16H11N3O2
277.28
MFCD04114290
ZYTWXMBGOUJDHJ-UHFFFAOYSA-N
2,2':6',2-terpyridine-4'-carboxylic acid, 2,2':6',2-terpyridine-4'-carboxylicacid, 2,6-bis pyridin-2-yl pyridine-4-carboxylic acid, 6-pyridin-2-yl-2,2'-bipyridine-4-carboxylic acid, acmc-1c3v7, 4'-carboxy-2,2':6',2-terpyridine, 2,2:6,2'-terpyridine-4-carboxylic acid, 1∼2∼,2∼2∼:2∼6∼,3∼2∼-terpyridine-2∼4∼-carboxylic acid, 2,2\\':6\\',2\\'\\'-terpyridine-4\\'-carboxylic acid
2762749
2,6-dipyridin-2-ylpyridine-4-carboxylic acid
OC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

2,2':6',2''-Terpyridine-4'-carboxylic acid is used as dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′:6′,2′′-Terpyridine-4′-carboxylic acid is used as dyestuff intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 280°C to 282°C
Color White to Cream
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 277.28
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2,2':6',2''-Terpyridine-4'-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'