missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2':5',2''-Terthiophene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2,2?:5?,2??-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2′:5′,2′′-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
2,2′:5′,2′′-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 89°C to 94°C |
Boiling Point | 160°C (0.1 mmHg) |
Quantity | 250 mg |
Beilstein | 178604 |
Merck Index | 14,9174 |
Solubility Information | Insoluble in water. |
Formula Weight | 248.39 |
Percent Purity | 99% |
Chemical Name or Material | 2,2':5',2''-Terthiophene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'