missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane, Environmental Grade, 99.5+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
Description
Suitable for pesticide residue analysis2,2,4-Trimethylpentane is used as a solvent in spectrophotometric analysis and in organic synthesis. Due to its high antiknock octane number, it is widely employed as a standard reference fuel for octane ratings. It acts as an entrainer in azeotropic distillation as well as thinner. It is involved as processing aids in the petroleum industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -107°C |
Density | 0.692 g/mL |
Boiling Point | 98°C to 99°C |
Flash Point | -7°C (19°F) |
Odor | Nearly odorless |
Refractive Index | 1.391 |
Quantity | 1 L |
UN Number | UN1262 |
Beilstein | 1696876 |
Merck Index | 14,5193 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'