missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals

Catalog Number p-216662
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
540-84-1
C8H18
114.232
MFCD00008943
NHTMVDHEPJAVLT-UHFFFAOYSA-N
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
540-84-1
C8H18
114.232
MFCD00008943
NHTMVDHEPJAVLT-UHFFFAOYSA-N
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY
Melting Point -107.0°C
Absorbance 0.01 max. at 250nm, 0.01 max. at 400nm, 0.04 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, 1 max. at 210nm
CAS Min % 98.5
CAS Max % 100.0
Density 0.6900g/mL
Boiling Point 98.0°C to 99.0°C
Quantity 500 mL
Flash Point −12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3915
Assay Percent Range 99%
Residue after Evaporation 0.001% max.
Specific Gravity 0.69
Linear Formula (CH3)2CHCH2C(CH3)3
Viscosity 0.51 mPa.s (22°C)
Beilstein 01, IV, 439
Water 0.02% max.
Merck Index 15, 5239
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol, soluble in benzene, toluene, xylene, chloroform,, dmf, ether, ccl4 and cs2, miscible with acetone, soluble in oils except castor oil
Formula Weight 114.23
Percent Purity 99%
Grade Spectroscopy
Chemical Name or Material 2, 2, 4-Trimethylpentane, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2,4-Trimethylpentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'