missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%, Thermo Scientific Chemicals

Catalog Number p-207495
Click to view available options
Quantity:
5 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
6846-50-0
C16H30O4
286.4
MFCD00059267
OMVSWZDEEGIJJI-UHFFFAOYSA-N
2,2,4-trimethyl-1,3-pentanediol diisobutyrate, kodaflex txib, 2,2,4-trimethylpentane-1,3-diyl bis 2-methylpropanoate, txib, propanoic acid, 2-methyl-, 2,2-dimethyl-1-1-methylethyl-1,3-propanediyl ester, 2,2,4-trimethylpentanediol-1,3-diisobutyrate, isobutyric acid, 1-isopropyl-2,2-dimethyltrimethylene ester, 1-isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-trimethyl-1,3-pentanediol ester, dsstox_cid_7635
23284
[2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate
CC(C)C(C(C)(C)COC(=O)C(C)C)OC(=O)C(C)C
This item is not returnable. View return policy
6846-50-0
C16H30O4
286.4
MFCD00059267
OMVSWZDEEGIJJI-UHFFFAOYSA-N
2,2,4-trimethyl-1,3-pentanediol diisobutyrate, kodaflex txib, 2,2,4-trimethylpentane-1,3-diyl bis 2-methylpropanoate, txib, propanoic acid, 2-methyl-, 2,2-dimethyl-1-1-methylethyl-1,3-propanediyl ester, 2,2,4-trimethylpentanediol-1,3-diisobutyrate, isobutyric acid, 1-isopropyl-2,2-dimethyltrimethylene ester, 1-isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-trimethyl-1,3-pentanediol ester, dsstox_cid_7635
23284
[2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate
CC(C)C(C(C)(C)COC(=O)C(C)C)OC(=O)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless
Density 0.9400g/mL
Boiling Point 280.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 5 g
Merck Index 15, 9399
Specific Gravity 0.94
Solubility Information Solubility in water: insoluble
Formula Weight 286.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
Show More Show Less
Product Identifier
  • 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'