missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2,4-Dichlorophenyl)ethanol, 97%

Brand:  Alfa Aesar™ H64345.03

Code : NEW

Additional Details : CAS Number : 81156-68-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15431509

  • £16.96 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(2,4-Dichlorophenyl)ethanol
191.05
6775864
81156-68-5
MFCD00192010
2,4-Dichlorophenethyl alcohol

Specifications

2-(2,4-Dichlorophenyl)ethanol
119°C (0.8mmHg) (lit.)
81156-68-5
Liquid
191.05
6775864
1.321g/mL
1g
1.5660
97%
MFCD00192010
2,4-Dichlorophenethyl alcohol
Videos
SDS