missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-[2-(4-[alpha-(4-Chlorophenyl)benzyl]-1-piperazinyl)ethoxy]acetamide, 97%

Brand:  Alfa Aesar™ H63408.03

Code : NEW

Additional Details : CAS Number : 83881-37-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478296

  • £226.84 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

2-[2-(4-[alpha-(4-Chlorophenyl)benzyl]-1-piperazinyl)ethoxy]acetamide
387.9
2-[2-(4-[4-Chlorophenyl(phenyl)methyl]-1-piperazinyl)ethoxy]acetamide
83881-37-2
MFCD01751128

Specifications

2-[2-(4-[alpha-(4-Chlorophenyl)benzyl]-1-piperazinyl)ethoxy]acetamide
Solid
387.9
White
1g
83881-37-2
97%
MFCD01751128
2-[2-(4-[4-Chlorophenyl(phenyl)methyl]-1-piperazinyl)ethoxy]acetamide
Videos
SDS