missing translation for 'onlineSavingsMsg'
Learn More

2,2,3-Trimethylbutane, 99%, Thermo Scientific Chemicals

Catalog Number 10685125
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
464-06-2
C7H16
100.2
MFCD00039846
ZISSAWUMDACLOM-UHFFFAOYSA-N
triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl
10044
2,2,3-trimethylbutane
CC(C)C(C)(C)C
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
464-06-2
C7H16
100.2
MFCD00039846
ZISSAWUMDACLOM-UHFFFAOYSA-N
triptan, triptane, butane, 2,2,3-trimethyl, unii-40v943jdgr, 2,3-trimethylbutane, butane,2,3-trimethyl, 2,2,3-trimethyl-butane, acmc-1ahi7, butane,2,2,3-trimethyl, 2,3,3-trimethylbutan-2-yl
10044
2,2,3-trimethylbutane
CC(C)C(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -25.0°C
Color Colorless
Density 0.6900g/mL
Boiling Point 81.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(CH3)2CH(CH3)2
Refractive Index 1.3880 to 1.3900 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.69
Formula Weight 100.2
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2, 2, 3-Trimethylbutane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2,3-Trimethylbutane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'